VWR® Modular Heating Blocks, Solid
Solid Block, Double, Double, 15.1 x 9.5 x 5.7 cm (6 x 33/4 x 21/4")
13259-297
Constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
For use as a low-temperature hotplate or for drilling a customized block. Available in single or double block size.
- Désignation:Solid Block, Double
- L×W×H:15.1 x 9.5 x 5.7 cm (6 x 33/4 x 21/4")
- Block Size:Double