Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sliding Track Curtains, Trax | Avantor

Sliding Track Curtains, Trax

8 ft., 6 ft.
Catalog # 12999-044
Fournisseur #:  STK20ST86
Sliding Track Curtains, Trax
Catalog # 12999-044
Sliding Track Curtains, Trax

8 ft., 6 ft.

Price
$1,599.55
Log in pour la disponibilité
Each
Product Details & Documents

These solid yet flexible standard curtains are designed for use as temporary or movable barriers

Shipping weights vary depending on size. Thickness: 20mil.

Ordering information: Includes a complete sliding track hardware mounting assembly.

Spécifications
  • Longueur:
    6 ft.
  • Width:
    8 ft.