OIML Class E2 Precision Weight Sets, Troemner | Avantor
OIML Class E2 Precision Weight Sets, Troemner
16, 5 kg - 1 g
Catalog # 12766-196
Supplier:   Troemner
Fournisseur #:  80780400
OIML Class E2 Precision Weight Sets, Troemner
Catalog # 12766-196
Supplier:  Troemner
OIML Class E2 Precision Weight Sets, Troemner
OIML Class E2 Precision Weight Sets, Troemner
16, 5 kg - 1 g
Price
$15,331.08
Log in pour la disponibilité
Each
Product Details & Documents
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are one-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.
 : VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWR[email protected] for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
Spécifications
  • Certification:
    NVLAP
  • Weight:
    5 kg - 1 g
  • No. of Weights:
    16