OIML Class E2 Precision Weights, Troemner | Avantor
OIML Class E2 Precision Weights, Troemner
10 g
Catalog # 12766-178
Product available on GSA Advantage®
Supplier:   Troemner
Fournisseur #:  80780296
OIML Class E2 Precision Weights, Troemner
Catalog # 12766-178
Supplier:  Troemner
Product available on GSA Advantage®
OIML Class E2 Precision Weights, Troemner
OIML Class E2 Precision Weights, Troemner
10 g
Price
459,09 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are one-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWR[email protected] for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
Spécifications
  • Certification:
    NVLAP
  • Weight:
    10 g
  • Cat. No.:
    12766-178