Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Modular Heating Blocks for Vials | Avantor

VWR® Modular Heating Blocks for Vials

19 mm Vial Block, Vials, Ext.Ø 19 mm, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2"), 12
Catalog # 12621-130
Supplier:   VWR International
VWR® Modular Heating Blocks for Vials
Catalog # 12621-130
Supplier:  VWR International
VWR® Modular Heating Blocks for Vials
VWR® Modular Heating Blocks for Vials

19 mm Vial Block, Vials, Ext.Ø 19 mm, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2"), 12

Price
$434.30
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

Designed for sample/serum and scintillation vials, these units are constructed from a solid anodized aluminum block.

  • Now with Best-in-Class, Exclusive 5-Year Warranty

The close contact of tubes to block walls allows for maximum heat retention. Single block size.

Spécifications
  • Désignation:
    19 mm Vial Block
  • For Vials:
    Ext.Ø 19 mm
  • L×W×H:
    9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
  • For Use With:
    Vials
  • No. of Wells:
    12