Infecon® Absorbent Strips, Com-Pac
7.6×15.2 cm (3×6"), 100 ml
Catalog # 11217-526
Supplier: Com-Pac
Supplier #: INF-1003
Jump to:
Product Details & Documents
Designed for use with Infecon® shippers and diagnostic packaging, these absorbent strips provide fast and reliable protection against spills.
Specifications
- L×l:7.6×15.2 cm (3×6")
- Capacité d'absorption:100 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



