Dissolution Calibration Tools, Quality Lab Accessories
Wobble Meter Includes Cover Style Bracket, Multi-Instrument
  11010-536
 :   QLA
 :  WOBMET-MI
Dissolution Calibration Tools, Quality Lab Accessories
  11010-536
 :  QLA
Dissolution Calibration Tools, Quality Lab Accessories
Dissolution Calibration Tools, Quality Lab Accessories
Wobble Meter Includes Cover Style Bracket, Multi-Instrument
$1,067.54
Each
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
  • Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
 : All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.
  • Désignation:
    Wobble Meter Includes Cover Style Bracket, Multi-Instrument