Miscellaneous Dissolution Accessories, Quality Lab Accessories
Plain Paper Rolls for Legacy VanKel Report Center II (Pack/25)
11010-402
: QLA
: PAP100-01
Everclear Water Bath Preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths.Use with DEIONIZED water
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, Rotating Bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE Suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.
- Désignation:Plain Paper Rolls for Legacy VanKel Report Center II (Pack/25)