Dissolution Vessels and Centering System Accessories, Quality Lab Accessories | Avantor
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Clear Glass Vessel for Agilent/VanKel/Varian, 500 ml
Catalog # 11010-332
Supplier:   QLA
Fournisseur #:  GLA500-01
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Catalog # 11010-332
Supplier:  QLA
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Clear Glass Vessel for Agilent/VanKel/Varian, 500 ml
Price
$285.58
Log in pour la disponibilité
Each
Product Details & Documents
QLA manufactures a wide selection of Precision Molded and Standard Glass Vessels for most dissolution testers.
  • All glass vessels are made of borosilicate glass in the USA
  • Vessels are available in clear and amber
  • UltraCenter Precision, Standard and Apex Vessels available with both glass and plastic rims
Good dissolution vessels are critical in obtaining correct and consistent results
UltraCenter Precision Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results. They are compatible with most dissolution baths. QLA's molded Apex vessels are patent pending.
We have the solution if you are having problems with vessel centering. The QLA AccuCenter Bracket provides ultimate centering accuracy on most dissolution baths (both old and new). It is adjustable and designed to center the vessel from the inside diameter and not the outside. When used with our UltaCenter Precision Vessels with glass or plastic rims, extreme centering accuracy can be achieved. Note: Standard glass vessels cannot be used in the AccuCenter Bracket.
Plastic Rimmed Vessels were designed to help minimize glass rim breakage that can occur in the lab. All QLA plastic rim vessels are dishwasher safe.
All vessels meet the physical dimensions of the USP and have a hemispherical bottom which should be perfectly rounded with no obvious defects. QLA glass vessels are acid etched, or laser marked on plastic rim, with a unique serial number and come with a Certificate of Conformance (COC), where applicable.
Certifications: All vessels are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an extra cost.
Spécifications
  • Volume:
    500 ml
  • Désignation:
    Clear Glass Vessel for Agilent/VanKel/Varian
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vessel Centering Bracket (EaseAlign) with 6 Replaceable Pins for Legacy VanKel Baths
    Vessel Centering Bracket (EaseAlign) with 6 Replaceable Pins for Legacy VanKel Baths
    11010-242
    Log in pour la disponibilité
    Each - $174.11
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vessel Centering Bracket (EaseAlign) Repair Kit (Set/12)
    Vessel Centering Bracket (EaseAlign) Repair Kit (Set/12)
    11010-244
    Log in pour la disponibilité
    Each - $78.19
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vessel Centering Bracket Peg
    Vessel Centering Bracket Peg
    11010-246
    Log in pour la disponibilité
    Each - $42.69
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Zymark/MultiDose Centering Ring
    Zymark/MultiDose Centering Ring
    11010-170
    Log in pour la disponibilité
    Each - $19.72
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Acculign Plastic Vessel Centering Ring for Distek Baths
    Acculign Plastic Vessel Centering Ring for Distek Baths
    11010-460
    Log in pour la disponibilité
    Each - $33.17
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Zymark/Multidose Vessel Retainer
    Zymark/Multidose Vessel Retainer
    11010-458
    Log in pour la disponibilité
    Each - $220.16
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vessel Retainer Assembly for Distek 2100 and 6300 baths
    Vessel Retainer Assembly for Distek 2100 and 6300 baths
    11010-510
    Log in pour la disponibilité
    Each - $19.49
Informations sur le produit
  • Catalog #
    Availability
    Price
  • AccuCenter Bracket for Distek 2100 Baths includes hardware
    AccuCenter Bracket for Distek 2100 Baths includes hardware
    75849-090
    Log in pour la disponibilité
    Each - $276.50
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vessel Clamp Knob for 708-DS baths
    Vessel Clamp Knob for 708-DS baths
    75848-944
    Log in pour la disponibilité
    Pack of 2 - $82.93
Informations sur le produit
  • Catalog #
    Availability
    Price
  • AccuCenter Bracket for Distek 2500 Baths includes hardware
    AccuCenter Bracket for Distek 2500 Baths includes hardware
    75849-092
    Log in pour la disponibilité
    Each - $287.16
Informations sur le produit
  • Catalog #
    Availability
    Price
  • AccuCenter Bracket for Hanson SR8-Plus Baths includes hardware
    AccuCenter Bracket for Hanson SR8-Plus Baths includes hardware
    75849-096
    Log in pour la disponibilité
    Each - $287.16
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vessel Hold Down Ring for UltraCenter Vessels on Erweka DT Baths
    Vessel Hold Down Ring for UltraCenter Vessels on Erweka DT Baths
    75849-266
    Log in pour la disponibilité
    Each - $135.61
Informations sur le produit
  • Catalog #
    Availability
    Price
  • AccuCenter Bracket for Legacy VanKel and Agilent 708-DS Baths includes hardware
    AccuCenter Bracket for Legacy VanKel and Agilent 708-DS Baths includes hardware
    75849-098
    Log in pour la disponibilité
    Each - $287.16