Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Product Details & Documents
QLA manufactures a wide selection of Precision Molded and Standard Glass Vessels for most dissolution testers.
- All glass vessels are made of borosilicate glass in the USA
- Vessels are available in clear and amber
- UltraCenter Precision, Standard and Apex Vessels available with both glass and plastic rims
Good dissolution vessels are critical in obtaining correct and consistent results
UltraCenter Precision Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results. They are compatible with most dissolution baths. QLA's molded Apex vessels are patent pending.
We have the solution if you are having problems with vessel centering. The QLA AccuCenter Bracket provides ultimate centering accuracy on most dissolution baths (both old and new). It is adjustable and designed to center the vessel from the inside diameter and not the outside. When used with our UltaCenter Precision Vessels with glass or plastic rims, extreme centering accuracy can be achieved. Note: Standard glass vessels cannot be used in the AccuCenter Bracket.
Plastic Rimmed Vessels were designed to help minimize glass rim breakage that can occur in the lab. All QLA plastic rim vessels are dishwasher safe.
All vessels meet the physical dimensions of the USP and have a hemispherical bottom which should be perfectly rounded with no obvious defects. QLA glass vessels are acid etched, or laser marked on plastic rim, with a unique serial number and come with a Certificate of Conformance (COC), where applicable.
Certifications: All vessels are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an extra cost.
Specifications
- Volume:2000 ml
- Désignation:Clear Glass Vessel for Agilent/VanKel/Varian, Serialized
Accessories
Product Information
- Catalog #AvailabilityPrice
- Vessel Centering Bracket (EaseAlign) with 6 Replaceable Pins for Legacy VanKel BathsVessel Centering Bracket (EaseAlign) with 6 Replaceable Pins for Legacy VanKel Baths11010-242Log in for availabilityEach - $174.11
Product Information
- Catalog #AvailabilityPrice
- Vessel Centering Bracket (EaseAlign) Repair Kit (Set/12)Vessel Centering Bracket (EaseAlign) Repair Kit (Set/12)11010-244Log in for availabilityEach - $78.19
Product Information
- Catalog #AvailabilityPrice
- Vessel Centering Bracket Peg
Product Information
- Catalog #AvailabilityPrice
- Zymark/MultiDose Centering Ring
Product Information
- Catalog #AvailabilityPrice
- Acculign Plastic Vessel Centering Ring for Distek BathsAcculign Plastic Vessel Centering Ring for Distek Baths11010-460Log in for availabilityEach - $33.17
Product Information
- Catalog #AvailabilityPrice
- Zymark/Multidose Vessel Retainer
Product Information
- Catalog #AvailabilityPrice
- Vessel Retainer Assembly for Distek 2100 and 6300 bathsVessel Retainer Assembly for Distek 2100 and 6300 baths11010-510Log in for availabilityEach - $19.49
Product Information
- Catalog #AvailabilityPrice
- AccuCenter Bracket for Distek 2100 Baths includes hardwareAccuCenter Bracket for Distek 2100 Baths includes hardware75849-090Log in for availabilityEach - $276.50
Product Information
- Catalog #AvailabilityPrice
- Vessel Clamp Knob for 708-DS baths
Product Information
- Catalog #AvailabilityPrice
- AccuCenter Bracket for Distek 2500 Baths includes hardwareAccuCenter Bracket for Distek 2500 Baths includes hardware75849-092Log in for availabilityEach - $287.16
Product Information
- Catalog #AvailabilityPrice
- AccuCenter Bracket for Hanson SR8-Plus Baths includes hardwareAccuCenter Bracket for Hanson SR8-Plus Baths includes hardware75849-096Log in for availabilityEach - $287.16
Product Information
- Catalog #AvailabilityPrice
- Vessel Hold Down Ring for UltraCenter Vessels on Erweka DT BathsVessel Hold Down Ring for UltraCenter Vessels on Erweka DT Baths75849-266Log in for availabilityEach - $135.61
Product Information
- Catalog #AvailabilityPrice
- AccuCenter Bracket for Legacy VanKel and Agilent 708-DS Baths includes hardwareAccuCenter Bracket for Legacy VanKel and Agilent 708-DS Baths includes hardware75849-098Log in for availabilityEach - $287.16