Disintegration Testing, Quality Lab Accessories
Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
Catalog # 11010-224
Supplier: QLA
Fournisseur #: DISTUB-P6
Jump to:
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
- Désignation:Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm