Disintegration Testing, Quality Lab Accessories | Avantor
Disintegration Testing, Quality Lab Accessories
Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
Catalog # 11010-224
Supplier:   QLA
Fournisseur #:  DISTUB-P6
Disintegration Testing, Quality Lab Accessories
Catalog # 11010-224
Supplier:  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
Price
$646.06
Log in pour la disponibilité
Each
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
  • Désignation:
    Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm