Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
Catalog # 11010-222
Supplier:   QLA
Supplier #:  DISTUB-06
Disintegration Testing, Quality Lab Accessories
Catalog # 11010-222
Supplier:  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
Price
$180.16
Log in pour la disponibilité
Pack of 6
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
  • Désignation:
    Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm