Disintegration Testing, Quality Lab Accessories
Disintegration Basket Assembly with 6 Plastic Tubes and 10 Mesh SS Screens for VanKel Testers
Catalog # 11010-220
Supplier: QLA
Fournisseur #: DISASY-P6
Jump to:
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
- Désignation:Disintegration Basket Assembly with 6 Plastic Tubes and 10 Mesh SS Screens for VanKel Testers