Dissolution Calibration Tools, Quality Lab Accessories | Avantor
Dissolution Calibration Tools, Quality Lab Accessories
25 mm Depth Set Gauge, Stainless Steel, VanKel Style
Catalog # 11010-214
Supplier:   QLA
Fournisseur #:  DEPGAG-VK
Dissolution Calibration Tools, Quality Lab Accessories
Catalog # 11010-214
Supplier:  QLA
Dissolution Calibration Tools, Quality Lab Accessories
Dissolution Calibration Tools, Quality Lab Accessories
25 mm Depth Set Gauge, Stainless Steel, VanKel Style
Price
$134.30
Log in pour la disponibilité
Each
Product Details & Documents
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
  • Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
Certifications: All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.
Spécifications
  • Désignation:
    25 mm Depth Set Gauge, Stainless Steel, VanKel Style