Dissolution Calibration Tools, Quality Lab Accessories
25 mm Depth Set Gauge, Stainless Steel, VanKel Style
Catalog # 11010-214
Supplier: QLA
Fournisseur #: DEPGAG-VK
Jump to:
Product Details & Documents
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
- Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
Certifications: All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.
Spécifications
- Désignation:25 mm Depth Set Gauge, Stainless Steel, VanKel Style
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.