Tris-HCl Buffer, 1.5 M Sterile Solution
Tris-HCl Buffer 1.5 M, 1 L, 8.8
Catalog # CA77442-930
New Product
Supplier: Teknova
Fournisseur #: T1588
Tris-HCl Buffer, 1.5 M Sterile Solution
Catalog # CA77442-930
Supplier: Teknova
New Product
CAS Number:
Jump to:
Product Details & Documents
1.5 M Tris (hydroxymethyl) Aminomethane solution adjusted to pH 8.8.
Spécifications
Spécifications
- Désignation:Tris-HCl Buffer 1.5 M
- Taille:1 L
- pH:8.8
- Cat. No.:CA77442-930
- Storage temperature:Store at room temperature. Shipped at ambient temperature.



