Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
1M Sodium Phosphate Buffer

1M Sodium Phosphate Buffer

1M sodium phosphate buffer, 1000 ml, Plastic bottle, 7.2
Catalog # CA77442-715
New Product
Supplier:   Teknova
Supplier #:  P3072
1M Sodium Phosphate Buffer
Catalog # CA77442-715
Supplier:  Teknova
New Product
CAS Number:  
1M Sodium Phosphate Buffer

1M sodium phosphate buffer, 1000 ml, Plastic bottle, 7.2

CAS Number:  
Price
$73.99
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

312 mM sodium phosphate monobasic, 688 mM sodium phosphate dibasic.

Specifications

Specifications

  • Type de conditionnement:
    Plastic bottle
  • Désignation:
    1M sodium phosphate buffer
  • Taille:
    1000 ml
  • pH:
    7.2
  • Cat. No.:
    CA77442-715
  • Sterility:
    Sterile

Specifications