solution 100 mg/mL
1 ml
CA77528-960
New Product
: Teknova
: C8001
Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.
It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.
- :4697-36-3
- :C₁₇H₁₈N₂O₆S
- Taille:1 ml