solution 100 mg/mL
1 ml
  CA77528-960
New Product
 :   Teknova
 :  C8001
solution 100 mg/mL
  CA77528-960
 :  Teknova
New Product
 :  4697-36-3
solution 100 mg/mL
1 ml
 :  4697-36-3
$564.66
Each
Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.
It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.
  •  :4697-36-3
  •  :C₁₇H₁₈N₂O₆S
  • Taille:
    1 ml