Eyepieces for Microscope
Wide field 15x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 15
Catalog # 78620-748
New Product
Supplier:   Aven Tools
 :  26800B-449
Eyepieces for Microscope
Catalog # 78620-748
Supplier:  Aven Tools
New Product
Eyepieces for Microscope
Wide field 15x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 15
Price
552,68 $
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Désignation:
    Wide field 15x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 15