Eyepieces for Microscope
Eyepieces for Microscope
Wide field 10x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 23
Catalog # 78620-614
New Product
Supplier:   Aven Tools
Supplier #:  26800B-448
Eyepieces for Microscope
Catalog # 78620-614
Supplier:  Aven Tools
New Product
Eyepieces for Microscope
Wide field 10x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 23
Price
$442.15
Log in for availability
Each
Product Details & Documents
Specifications
  • Désignation:
    Wide field 10x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 23