Eyepieces for Microscope | Avantor
Eyepieces for Microscope
10x eyepiece with 5:100 mm scale reticle for SPZ microscopes
Catalog # 78620-882
New Product
Supplier:   Aven Tools
Fournisseur #:  26800B-456
Eyepieces for Microscope
Catalog # 78620-882
Supplier:  Aven Tools
New Product
Eyepieces for Microscope
10x eyepiece with 5:100 mm scale reticle for SPZ microscopes
Price
$962.88
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Désignation:
    10x eyepiece with 5:100 mm scale reticle for SPZ microscopes