Eyepieces for Microscope
Eyepieces for Microscope
Wide field 20x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 11.4
Catalog # 78621-336
New Product
Supplier:   Aven Tools
Supplier #:  26800B-443
Eyepieces for Microscope
Catalog # 78621-336
Supplier:  Aven Tools
New Product
Eyepieces for Microscope
Wide field 20x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 11.4
Price
$552.68
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Désignation:
    Wide field 20x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 11.4