Eyepieces for Microscope
Wide field 20x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 11.4
Catalog # 78621-336
New Product
Supplier: Aven Tools
Supplier #: 26800B-443
Jump to:
Product Details & Documents
Spécifications
- Désignation:Wide field 20x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 11.4