Eyepieces for Microscope
Eyepieces for Microscope
Wide field 10x eyepiece for DSZ-44 and NSW microscopes, Ø 29.95 mm, FN: 23
  78621-338
New Product
 :   Aven Tools
Supplier #:  26800B-441
Eyepieces for Microscope
  78621-338
New Product
Eyepieces for Microscope
Wide field 10x eyepiece for DSZ-44 and NSW microscopes, Ø 29.95 mm, FN: 23
$442.15
Each
  • Désignation:
    Wide field 10x eyepiece for DSZ-44 and NSW microscopes, Ø 29.95 mm, FN: 23