Computar MLH 10× Lens
Adapter plate for Macro lens
Catalog # 78621-380
New Product
Supplier: Aven Tools
Fournisseur #: 26700-181AP
Jump to:
Product Details & Documents
Computar manual zoom lenses provide manual adjustment of focal length, iris and focus.
- f/5.6 to 32C manual iris
- 21.6 to 4.5° diagonal FOV
- Manual zoom (10×) and focus
- Focus: 0.15 to 0.45 m
- C-Mount connection
For use where a variable angle of view is required. Main applications for this lens include inspection, machine vision and robotics.This system requires an adapter plate.
Spécifications
- Désignation:Adapter plate for Macro lens