Computar MLH 10× Lens | Avantor
Computar MLH 10× Lens
Adapter plate for Macro lens
Catalog # 78621-380
New Product
Supplier:   Aven Tools
Fournisseur #:  26700-181AP
Computar MLH 10× Lens
Catalog # 78621-380
Supplier:  Aven Tools
New Product
Computar MLH 10× Lens
Adapter plate for Macro lens
Price
$175.85
Log in pour la disponibilité
Each
Product Details & Documents
Computar manual zoom lenses provide manual adjustment of focal length, iris and focus.
  • f/5.6 to 32C manual iris
  • 21.6 to 4.5° diagonal FOV
  • Manual zoom (10×) and focus
  • Focus: 0.15 to 0.45 m
  • C-Mount connection
For use where a variable angle of view is required. Main applications for this lens include inspection, machine vision and robotics.This system requires an adapter plate.
Spécifications
  • Désignation:
    Adapter plate for Macro lens