Accessories for SharpVue Microscopes
10 diopter lens for Sharp Vue XT; Magnification: 4.7 - 94× (based on a 23.5" monitor)
78620-524
New Product
Supplier #: 26700-137-10D
- Désignation:10 diopter lens for Sharp Vue XT; Magnification: 4.7 - 94× (based on a 23.5" monitor)