Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Professional Automatic Wire Strippers | Avantor

Professional Automatic Wire Strippers

Professional automatic wire stripper, Range: 18 - 8 AWG, Carbon steel, 18/14/10/8 AWG wire stripping, Spring loaded, 0.4536 kg
Catalog # 78621-392
New Product
Supplier:   Aven Tools
Fournisseur #:  10105B
Professional Automatic Wire Strippers
Catalog # 78621-392
Supplier:  Aven Tools
New Product
Professional Automatic Wire Strippers

Professional automatic wire stripper, Range: 18 - 8 AWG, Carbon steel, 18/14/10/8 AWG wire stripping, Spring loaded, 0.4536 kg

Price
$67.02
Log in pour la disponibilité
Each
Product Details & Documents

Professional grade wire stripper with a spring-loaded handle and a comfort grip, designed for easy wire stripping. Includes a built-in safety shield.

  • Quick and easy wire stripping

    Professional-quality tool for fast & consistent wire stripping

    Easy to use - simply insert wire and squeeze handle

    Spring mechanism automatically holds wire, cuts & strips insulation clean leaving bare wire

    Non-slip grips, steel mechanism - great for home, auto, A/V wiring

    Built-in peg hole for storing on a pegboard, and can be used to hang from a tool belt

    Tempered high-carbon steel blades are designed for thousands of cuts

Spécifications
  • Désignation:
    Professional automatic wire stripper, Range: 18 - 8 AWG
  • Matière:
    Carbon steel
  • Cutting Capacity:
    18/14/10/8 AWG wire stripping
  • Weight:
    0.4536 kg
  • Handle Type:
    Spring loaded
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.