Nitrate Test Kits | Avantor
Nitrate Test Kits
NED, 0 - 1.5 ppm, Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff, 30
Catalog # 78620-296
New Product
Supplier:   CHEMetrics
Fournisseur #:  K-6973
Nitrate Test Kits
Catalog # 78620-296
Supplier:  CHEMetrics
New Product
CAS Number:  
Nitrate Test Kits
Nitrate Test Kits
NED, 0 - 1.5 ppm, Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff, 30
CAS Number:  
Price
$110.30
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The NED method is widely used in the quantitative analysis of nitrite and nitrate in water samples. NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, lower detection limits and measurement ranges are possible vs. chromotropic acid kits. In addition, the nitrate measurement uses only zinc which is less toxic than cadmium, making handling and disposal safer.
  • Measurement via zinc reduction only, safer handling and disposal
    Improved sensitivity and faster analysis time
    Simplified reordering, visual refills include NED, Zn, Acidifier and buffer under single catalog code
CHEMetrics Nitrate Test Kits use the N-(1-Naphthyl) ethylenediamine (NED) method to measure nitrate in potable, surface, storm, ground, sea, and wastewater. These kits offer improved sensitivity and faster analysis time (10 vs. 13 min) compared to our current chromotropic acid-based kits.
Like our legacy chromotropic acid reagent, NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, we are now able to offer lower detection limits and measurement ranges than our chromotropic acid kits provide.
Spécifications
Spécifications
  • Méthode chimique:
    NED
  • Utilisation:
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
  • Range:
    0 - 1.5 ppm
  • No. of Tests:
    30
Spécifications
Certains produits peuvent apparaître comme restreints
Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.
Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)
Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.