NEBNext® Enzymatic Methyl-seq v2 Conversion Module
NEBNext® Enzymatic Methyl-seq v2 Conversion Module
NEBNext® enzymatic methyl-seq v2 conversion module, 24 reactions
Catalog # CA77938-738
New Product
Supplier #:  E8020S
NEBNext® Enzymatic Methyl-seq v2 Conversion Module
Catalog # CA77938-738
New Product
CAS Number:  
NEBNext® Enzymatic Methyl-seq v2 Conversion Module
NEBNext® Enzymatic Methyl-seq v2 Conversion Module
NEBNext® enzymatic methyl-seq v2 conversion module, 24 reactions
CAS Number:  
Price
$279.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
NEBNext Enzymatic Methyl-seq (EM-seq) is a high-performance enzyme-based alternative to bisulfite conversion for the identification of 5mC and 5hmC.
  • High efficiency conversion
  • 0.1 to 200 ng input range
  • Minimal DNA damage
  • Minimized GC bias
  • 5mC and 5hmC detection
Unlike bisulfite conversion, this highly efficient method minimizes DNA damage, resulting in superior detection of methylated cytosines, with fewer sequencing reads. The new NEBNext Enzymatic Methyl-seq v2 Conversion Module has a wider input range (as low as 100 pg) than the original EM-seq Conversion Module.
For use in Illumina sequencing workflows, we recommend use of the NEBNext Enzymatic Methyl-seq v2 Kit instead of this module. The full kit includes the EM-seq Adaptor, which has been optimized for high performance in the EM-seq workflow, as well as the high-efficiency NEBNext Ultra II library prep reagents, and these components are not included in the Conversion Module.
Specifications
Specifications
  • Désignation:
    NEBNext® enzymatic methyl-seq v2 conversion module
  • Taille:
    24 reactions
Specifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.