Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Transferpette® pro, Ejector Set (upper and lower) | Avantor

Transferpette® pro, Ejector Set (upper and lower)

0.25 - 2.5 mL
Catalog # 78599-796
New Product
Supplier:   Brandtech
Fournisseur #:  704755
Transferpette® pro, Ejector Set (upper and lower)
Catalog # 78599-796
Supplier:  Brandtech
New Product
Transferpette® pro, Ejector Set (upper and lower)
Transferpette® pro, Ejector Set (upper and lower)

0.25 - 2.5 mL

Price
$76.55
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

Replacement ejector set.

  • Ejector set (upper and lower part) for use with Transferpette® pro single channel pipettors, 2.5 to 10ml

Designed for a perfect fit, simple installation, and long-lasting reliability.

Spécifications
  • Volume:
    0.25 - 2.5 mL
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.