Transferpette® pro, Ejector (bottom part)
20 - 200 µl
78599-700
New Product
: 704713
Replacement lower ejector.
Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
- Volume:20 - 200 µl