Transferpette® pro, Ejector (bottom part)
5 - 50 µl
78599-748
New Product
Supplier #: 704711
Replacement lower ejector.
Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
- Volume:5 - 50 µl