Transferpette® pro, Ejector (bottom part)
0.5 - 10 µl
  78599-706
New Product
 :   Brandtech
 :  704707
Transferpette® pro, Ejector (bottom part)
  78599-706
 :  Brandtech
New Product
Transferpette® pro, Ejector (bottom part)
Transferpette® pro, Ejector (bottom part)
0.5 - 10 µl
$43.83
Each
Replacement lower ejector.
  • Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
  • Volume:
    0.5 - 10 µl