Transferpette® pro, Ejector (bottom part)
0.1 - 2.5 µl
Catalog # 78599-712
New Product
Supplier: Brandtech
: 704706
Jump to:
Product Details & Documents
Replacement lower ejector.
Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
Specifications
- Volume:0.1 - 2.5 µl