Transferpette® pro, Ejector (bottom part) | Avantor
Transferpette® pro, Ejector (bottom part)
10 - 100 µl
Catalog # 78599-754
New Product
Supplier:   Brandtech
Fournisseur #:  704712
Transferpette® pro, Ejector (bottom part)
Catalog # 78599-754
Supplier:  Brandtech
New Product
Transferpette® pro, Ejector (bottom part)
Transferpette® pro, Ejector (bottom part)
10 - 100 µl
Price
$43.83
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Replacement lower ejector.
  • Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
Spécifications
  • Volume:
    10 - 100 µl