Instant Explosive Detection Wipes
Instant Explosive Detection Wipes
Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)
Catalog # 78568-310
New Product
Supplier:   TRACE EYE D LLC
Supplier #:  948254
Instant Explosive Detection Wipes
Catalog # 78568-310
Supplier:  TRACE EYE D LLC
New Product
Instant Explosive Detection Wipes
Instant Explosive Detection Wipes
Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)
Price
$414.73
Log in pour la disponibilité
Pack of 50
Minimum order quantity is 1
Product Details & Documents
Trace Eye-D instant explosives detection wipes provide rapid, field-ready identification of trace explosive residues, including TNT, PETN, RDX, TATP, ammonium nitrate, chlorates, and perchlorates. They require no specialized equipment and use simple color change chemistry for safe on-scene screening.
  • Rapid detection results of trace explosive residues
  • Field-ready packaging for on-scene operations
  • Trusted globally by law enforcement, corrections, and security teams
  • Simple color change results; no instruments, mixing, or glass ampoules
  • Works on solids, liquids, packaging, and surfaces
  • Minimizes exposure with safe stand-off sampling
  • Compact, lightweight, and easy to carry in any kit
  • Durable 6×3" polyester towelette allows refolding for testing several areas with one wipe
  • Negative result can be disposed of in common waste stream
  • Made in the USA
Trace Eye-D instant explosives detection wipes are simple to use, and require no specialized equipment. They employ color change chemistry for fast on-scene screening; just open, wipe, and look. Each wipe features a large 6×3" 100% polyester towelette, refoldable to test multiple surfaces, and is housed in a durable 4×3" multi-layer pouch (foil, LDPE, polyester, EAA) weighing 2.54 g total. Trace Eye-D wipes are pretreated, and must be used within 30 minutes after opening. They should be used within one year of purchase for optimal results. Trace Eye-D wipes are unaffected by heat, cold, or altitude and are lightweight and portable, making them ideal for law enforcement, military, aviation security, corrections, K-9, and bomb squad personnel.
Chemical reaction attributes: Aliphatic explosives detection wipes utilize Griess. Chlorate-based explosives detection wipes utilize REDOX. Aromatic explosives detection wipes utilize Meisenheimer Complex Reaction/Formation. Peroxide-based explosives detection wipes utilize REDOX. Perchlorate-based Oxidizer detection wipes utilize Precipitation Reaction.
 : Berry compliant.
 : Resealable bag contains 10 individually wrapped wipes; bulk options are loose indivudually wrapped wipes (no outer resealable packaging).
Spécifications
  • Désignation:
    Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)