Fentanyl Test Strips, Harm Reduction | Avantor
Fentanyl Test Strips, Harm Reduction
Fentanyl test strips, 10 ng/ml, 25 tests
Catalog # 78535-044
New Product
Supplier:   W.H.P.M INC
Fournisseur #:  WFT-ST25
Fentanyl Test Strips, Harm Reduction
Catalog # 78535-044
Supplier:  W.H.P.M INC
New Product
Fentanyl Test Strips, Harm Reduction
Fentanyl Test Strips, Harm Reduction
Fentanyl test strips, 10 ng/ml, 25 tests
Price
$44.24
Log in pour la disponibilité
Box of 25
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Developed specifically for harm reduction use, WHPM’s fentanyl test strips have both high sensitivity and high selectivity, allowing them to detect even tiny amounts of fentanyl in drug samples without cross-reacting with non-fentanyl related substances.
  • 10 ng/ml cut-off concentration
  • Quick reference guide instructions
The WHPM fentanyl test strips were developed specifically for harm reduction use. With a detection cutoff of 10 ng/ml, they are one of the most sensitive test strips on the market. They are also highly selective to fentanyl and its analogs, meaning they are less likely to cross-react with other, non-fentanyl related drugs.
Fentanyl is a powerful synthetic opioid fifty times stronger than heroin. Since about 2018 it has become the dominant opioid on the illicit market. Found in pills and powders, including non-opioid drugs and counterfeit pharmaceuticals, fentanyl is responsible for tens of thousands of deaths a year in the U.S. alone.
Spécifications
  • Désignation:
    Fentanyl test strips
  • Taille:
    25 tests
  • Sensitivity:
    10 ng/ml