Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Solvant 2000

Solvant 2000

Solvant 2000, toluene and xylene substitute, 10 L, Jug
Catalog # CA78494-380
New Product
Supplier:   CHAPTEC INC CA
Supplier #:  STY6750-10
Solvant 2000
Catalog # CA78494-380
Supplier:  CHAPTEC INC CA
New Product
CAS Number:  
Solvant 2000
Solvant 2000

Solvant 2000, toluene and xylene substitute, 10 L, Jug

CAS Number:  
Price
$316.00
Log in for availability
Each(1/Itm/Bx)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Toluene and xylene substitute.

  • General-purpose organic solvent

  • Specially formulated to replace toluene or xylene in histology and cytology procedures
  • Miscible with alcohol and paraffin
  • Can be used in the clearing step during processing
  • Also used  in the deparaffinization of slides and preparation for mounting
  • Biodegradable and less harmful, it offers a higher flash point compared to standard aromatic hydrocarbons

Specifications

Specifications

  • Type de conditionnement:
    Jug
  • Désignation:
    Solvant 2000, toluene and xylene substitute
  • Volume:
    10 L
  • Cat. No.:
    CA78494-380
  • Form:
    Colorless liquid

Specifications