Stopper Assembly
Stopper assembly, #6 taper, 30 mm
Catalog # 78473-510
New Product
Supplier: SAVILLEX
: 730-0066
Jump to:
Product Details & Documents
For use in glass flasks
Ferrule nut accepts ³/₈" OD tube
Spécifications
- Désignation:Stopper assembly, #6 taper
- Taille:30 mm