Column Component Closures
Column component closure, with two ¹/₂" outer Ø tube port, 58 mm
Catalog # 78474-155
New Product
Supplier: SAVILLEX
Fournisseur #: 600-058-42
Jump to:
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Spécifications
- Désignation:Column component closure, with two ¹/₂" outer Ø tube port, 58 mm