Column Component Closures
Column component closure, 58 mm
78474-153
New Product
: SAVILLEX
: 600-058-06
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
- Désignation:Column component closure, 58 mm