Column Component Closures | Avantor
Column Component Closures
Column component closure, with two ¹/₈" push in transfer side ports, 24 mm
Catalog # 78473-842
New Product
Supplier:   SAVILLEX
Fournisseur #:  600-024-23
Column Component Closures
Catalog # 78473-842
Supplier:  SAVILLEX
New Product
Column Component Closures
Column component closure, with two ¹/₈" push in transfer side ports, 24 mm
Price
$56.90
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Spécifications
  • Désignation:
    Column component closure, with two ¹/₈" push in transfer side ports, 24 mm