Column Component Closures
Column component closure, with two ¹/₄" outer Ø tube port, 58 mm
Catalog # 78473-600
New Product
Supplier: SAVILLEX
Supplier #: 600-058-33
Jump to:
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Specifications
- Désignation:Column component closure, with two ¹/₄" outer Ø tube port, 58 mm