Column Component Closures | Avantor
Column Component Closures
Column component closure, with two ¹/₈" outer Ø tube port, 58 mm
Catalog # 78473-538
New Product
Supplier:   SAVILLEX
Fournisseur #:  600-058-24
Column Component Closures
Catalog # 78473-538
Supplier:  SAVILLEX
New Product
Column Component Closures
Column component closure, with two ¹/₈" outer Ø tube port, 58 mm
Price
$136.91
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Spécifications
  • Désignation:
    Column component closure, with two ¹/₈" outer Ø tube port, 58 mm