CRISPR Adaptive Immunity
CRISPR Adaptive Immunity
CRISPR adaptive immunity, 1
Catalog # 470361-114
New Product
Supplier #:  CSCIAK-01
CRISPR Adaptive Immunity
Catalog # 470361-114
New Product
CRISPR Adaptive Immunity
CRISPR Adaptive Immunity
CRISPR adaptive immunity, 1
Price
$109.02
Log in for availability
Kit of 1 Case of 8
Product Details & Documents
What does CRISPR do?
  • Interactive modeling
  • How CRISPR was discovered and what it does
  • Reusable
Discover how bacteria remember past viral infections and activate their CRISPR systems to search for and destroy the remembered virus’ DNA - preventing infection. This introduction to CRISPR biology will prepare your students to use making the cut with CRISPR-Cas9 and understand how the Cas9 protein works.
With the CRISPR Adaptive Immunity Kit's engaging foam pieces, your students can:
Build the bacteria’s CRISPR array which includes Cas9, trace, spacer and repeats.
Detect the presence of viral double-stranded DNA.
Integrate a short fragment of this viral genome into the bacteria’s CRISPR array.
Express the CRISPR array to create the Cas9 proteins and the guide RNA.
Specifications
  • Désignation:
    CRISPR adaptive immunity
  • No. of Lab Groups:
    1
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.