Mining Sampling Bags, Clear Poly
Poly sample bag, 4 mil, 8×14", Plastic, Clear
Catalog # 80701-958
New Product
Supplier: SELENE PLASTIC BAGS CA
Supplier #: 0814391000CL
Jump to:
Product Details & Documents
Clear poly, mining sampling bags in various sizes and mil thicknesses.
Suitable for field and lab applications
- Made in Canada
Side welded seal imparts improved, with greater strength, and a resulting solid tubing bottom edge.
Spécifications
- Color:Clear
- Désignation:Poly sample bag
- Dimensions:8×14"
- Matière:Plastic
- Épaisseur:4 mil