Microbial Count Plate
Vibrio parahaemolyticus count plate, Gray colony with halo, 36±1 °C, 18-24h incubation
Catalog # 78173-204
New Product
 :  MCPG02825
Microbial Count Plate
Catalog # 78173-204
New Product
CAS Number:  
Microbial Count Plate
Vibrio parahaemolyticus count plate, Gray colony with halo, 36±1 °C, 18-24h incubation
CAS Number:  
Price
$388.16
Log in pour la disponibilité
Pack of 25
Product Details & Documents
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Spécifications
Spécifications
  • Désignation:
    Vibrio parahaemolyticus count plate
  • Format:
    Gray colony with halo, 36±1 °C, 18-24h incubation
Spécifications