Microbial Count Plate
Microbial Count Plate
Aerobic bacillus count plate, Red colony, 21±1 °C, 48-60h incubation
Catalog # 78172-698
New Product
Supplier #:  MCPG02425
Microbial Count Plate
Catalog # 78172-698
New Product
CAS Number:  
Microbial Count Plate
Aerobic bacillus count plate, Red colony, 21±1 °C, 48-60h incubation
CAS Number:  
Price
$388.16
Log in for availability
Pack of 25
Product Details & Documents
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Specifications
Specifications
  • Désignation:
    Aerobic bacillus count plate
  • Format:
    Red colony, 21±1 °C, 48-60h incubation
Specifications