Scleroderma CD56+ NK Cells
Scleroderma CD56+ NK cells, Cryopreserved, Single donor, 1×10⁷ cells (Vial), Human, adult
Catalog # 78170-882
New Product
Supplier: SANGUINE BIOSCIENCES INC
Supplier #: SYS-CD56-10M
Scleroderma CD56+ NK Cells
Catalog # 78170-882
Supplier: SANGUINE BIOSCIENCES INC
New Product
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Isolated CD56+ NK cells from Scleroderma donors to enable cytotoxicity and activation studies.
Disease-relevant CD56+ NK cells
Cryopreserved; QC’d for viability
Suitable for cytotoxicity and activation assays
Documentation available
CD56+ NK cells isolated Scleroderma donors are cryopreserved for reliable performance in cytotoxicity, activation, and signaling studies. These cells support flow cytometry, co-culture models, and transcriptomic analyses of innate immune function.
Specifications
Specifications
- Format de cellule:Cryopreserved
- Donneur unique/donneurs regroupés:Single donor
- Désignation:Scleroderma CD56+ NK cells
- Taille:1×10⁷ cells (Vial)
- Source:Human, adult
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



