Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
COPD PBMC

COPD PBMC

COPD PBMC, Cryopreserved, Single donor, 5×10⁷ cells (Vial), Human, adult
Catalog # 78170-558
New Product
Supplier #:  CPD-LKLT-50M
COPD PBMC
Catalog # 78170-558
New Product
CAS Number:  
COPD PBMC

COPD PBMC, Cryopreserved, Single donor, 5×10⁷ cells (Vial), Human, adult

CAS Number:  
Price
$1,671.18
Log in pour la disponibilité
Each
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Cryopreserved peripheral blood mononuclear cells (PBMCs) from clinically confirmed COPD donors for immunology and translational research.

  • Clinically confirmed COPD donors

    High-viability cryopreservation

    Research-ready aliquots

    Supports flow, cytokine, and gene-expression assays

    Documentation available

COPD PBMC are collected under IRB oversight and processed to preserve cellular integrity and functional responses. The PBMC format is suitable for phenotyping, cytokine release assays, co-culture systems, and multi-omics workflows investigating immune mechanisms in copd.

Spécifications

Spécifications

  • Format de cellule:
    Cryopreserved
  • Donneur unique/donneurs regroupés:
    Single donor
  • Désignation:
    COPD PBMC
  • Taille:
    5×10⁷ cells (Vial)
  • Source:
    Human, adult

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.