COPD PBMC
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cryopreserved peripheral blood mononuclear cells (PBMCs) from clinically confirmed COPD donors for immunology and translational research.
Clinically confirmed COPD donors
High-viability cryopreservation
Research-ready aliquots
Supports flow, cytokine, and gene-expression assays
Documentation available
COPD PBMC are collected under IRB oversight and processed to preserve cellular integrity and functional responses. The PBMC format is suitable for phenotyping, cytokine release assays, co-culture systems, and multi-omics workflows investigating immune mechanisms in copd.
Specifications
Specifications
- Format de cellule:Cryopreserved
- Donneur unique/donneurs regroupés:Single donor
- Désignation:COPD PBMC
- Taille:1×10⁸ cells (Vial)
- Source:Human, adult
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



