Epic™ Basic Protection Cleanroom Frocks
Product Details & Documents
Latex-free, low-lint frock for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Features include plastic snaps, knit wrists, and knit collar. Frocks are designed without pockets.
- Constructed from lightweight, breathable polypropylene non-woven fabric, ensuring comfort and reducing lint generation
- Snap-front design for easy gowning and removal while maintaining coverage integrity
- Elastic wrists improve glove interface
- Latex-free and lint-free design reduces allergy risk and minimizes particle shedding in cleanroom environments
- Individually bagged and cleanroom-packed to maintain garment integrity during storage and handling
- Helps prevent particulate transfer from personnel to sensitive environments, supporting GMP and ISO cleanroom protocols
- Disposable construction eliminates laundering costs and simplifies inventory management
- Ideal for ISO Class 7–8 cleanrooms, support areas, and non-critical manufacturing processes
The Epic™ Basic Protection Cleanroom Frock is designed to provide reliable coverage and contamination control for controlled environments where exposure risks are minimal but cleanliness standards remain critical. This garment offers a practical solution for cleanroom support areas, ISO Class 7–8 zones, and non-hazardous tasks, balancing comfort, compliance, and cost efficiency.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
: Aligns with basic cleanroom garment guidelines for controlled environments.
: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
Spécifications
- Taille de confection:S
- Color:White
- Seam type:Bound
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Press studs / snaps
- Tensile strength:3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Weight:30 g/m²
- Tearing strength:16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Cat. No.:78150-614
- Cleanroom class:10000/ISO 7
- Garment application:Cleanroom
- Pockets:Without pockets
- Fabric:SPP
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:6856 g/m²/24 h
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



